Learn More
OHAUS™ Flex System Clamp
OHAUS™ Flex System Clamp is designed to securely grip and position a variety of laboratory glassware.
Brand:  OHAUS™ 30400016
 View more versions of this product
Product Code. 15782429
  • £264.00 / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
  • Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
  • Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flex System Clamp
Clamp, Specialty, Flex, CLS-FLEXLL
18.000 in.
0 to 19 mm
Nickel-plated
13 mm
457 mm