Learn More
Proteintech™ FlexAble CoraLite™ Plus 550 Antibody Labeling Kit for Rabbit IgG
This Antibody Labeling Kit conjugates CoraLite™ 550 into Rabbit IgG antibodies.
Brand:  Proteintech™ KFA002-50
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17791509
  • £335.00 / 50 reactions
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
FlexAble CoraLite™ Plus 550 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 550 in any buffer condition to rabbit IgG primary antibodies from any supplier.
  • One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
  • Label your rabbit IgG antibody in <10 min without any additional equipment
Specifications
Specifications
50 μL CoraLite™ 550 FlexLinker for Rabbit IgG, 100 μL FlexQuencher for Rabbit IgG, 500 μL FlexBuffer, Manual, Protocol Magnet
1 Year at -20°C or 6 Months at +4°C
IgG
50 x 0.5 μg Reactions
554 nm/570 nm