Learn More
Proteintech™ FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Rabbit IgG
This Antibody Labeling Kit conjugates CoraLite™ 750 into Rabbit IgG antibodies.
Brand: Proteintech™ KFA004-50
Additional Details : Weight : 0.00010kg
Description
FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 750 in any buffer condition to rabbit IgG primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your rabbit IgG antibody in <10 min without any additional equipment
Specifications
50 μL CoraLite™ Plus 750 FlexLinker for Rabbit IgG, 100 μL FlexQuencher for Rabbit IgG, 500 μL FlexBuffer, Manual, Protocol Magnet | |
1 Year at -20°C or 6 Months at +4°C, Avoid Light | |
IgG |
50 x 0.5 μg Reactions | |
755 nm/780 nm | |
Rabbit |