missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Flexelene™ 135 C Tubing

Use Flexelene 135 C for processes or lab work that require stringent certifications

Brand:  Cole-Parmer™ 96181-10

 View more versions of this product

Product Code. 11759629

  • £830.00 / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ADI- and DEHP-free for the highest level of purity in your process.
  • Flexelene 135C is an ideal tubing for pharmaceutical, biotech, life science, research, and general-purpose lab applications.
  • Translucent, with no plasticizers, latex, or vinyl acetate.
  • Flexelene 135 C is also animal-derived ingredient-free.
  • Flexible tubing made of non-allergenic materials.
  • Ultra-low extractables and leachables.
  • FDA compliant and meets USP Class VI requirements.
  • Flexelene 135 C includes only additives approved in accordance with EP 3.1.3 and 3.1.6.
  • Base resin conforms with EC directive 202/72/EC.
  • Designed with the needs of bioprocessing and the pharmaceutical industry in mind.
  • Sterilize: Autoclave, ethylene oxide, e-beam or gamma irradiation.
Specifications

Specifications

3A;NSF-51;Non-DEHP;Non-Phthalate;USP;ADCF;European Pharmacopoeia;FDA;NSF-61
Autoclavable
Flexelene
50 ft.
3.175 mm
-46°F
50 ft./Pk.
Translucent
275°F
Made with no plasticizers, latex, or vinyl acetate, Flexelene 135 C is also animal-derived ingredient-free. This flexible tubing is made of non-allergenic materials and offers ultra-low extractables and leachables.
12.8 mm
19.1 mm
15.2 m
Flexelene 135C is an ideal tubing for pharmaceutical, biotech, life science, research, and general-purpose lab applications.
-43°C
Autoclave, ethylene oxide, e-beam, or gamma irradiation.
17 psi
135°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers