Learn More
Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes
Optimize readings and temperature accuracy for a variety of applications with these probes. Cole-Parmer™ Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes are ideal for high-temperature measurements and can be used with liquids, gases and semisolids.
Brand:  Digi-Sense™ 08516-97
 View more versions of this product
Product Code. 11752683
  • £129.00 / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Optimize readings and temperature accuracy for a variety of applications with these probes. Cole-Parmer™ Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes are ideal for high-temperature measurements and can be used with liquids, gases and semisolids.
All insulated-wire probes include straight insulated cable with no handle. Fiberglass and ceramic insulated probes have exposed junctions; use for high-temperature applications. Exposed junction probes are not recommended for use with aqueous solutions (type J exposed-junction probes will rust when exposed to moisture).
  • Each probe comes with color coded ANSI mini-connectors: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Ceramic Fiber-Insulated Wire Probes for high-temperature measurements.
  • General-Purpose Probes are designed for use with liquids, gases and semisolids.
  • ABS handles provide a light weight design for ease of use along with durability
  • Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging
Specifications
Specifications
ANSI
Thermocouple Probe
Thermocouple Type: K
63.5 cm
-250°C to +1100°C
Industrial environments where fast accurate readings are required
25 in.