Learn More
Memmert™ Flexible Pt Temperature Sensor
For Use With: ICP750
Brand: Memmert™ ICP750H8
This item is not returnable.
View return policy
Description
- Flexible Pt100 temperature sensor, positioned flexibly in chamber or load, for local temperature measurement (up to 2 additional sensors are possible)
- The measured temperature can, if required, be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
Specifications
ICP750 | |
Temperature Sensor |
Product Content Correction
Product Title
Memmert™ Flexible Pt Temperature Sensor > For Use With: ICP750
Spot an opportunity for improvement?Share a Content Correction