Learn More
Memmert™ Flexible Pt100 For Chamber Or Load, Display And Atmocontrol Compatible, For Un160Pa
Flexible temperature sensor for positioning in chamber or in load. Memmert™ PT100 Temperature Sensor is designed with a 4-pin connector for external temperature recording.
Brand:  Memmert™ UN160paH8
Product Code. 16374367
  • £197.00 / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
Includes: 4-Pin Connector for Temperature Recording
Description
Description
  • Can be positioned in chamber or in load
  • Includes 4-pin connector, according to NAMUR NE 28, for external temperature recording (load temperature) max. 3 sensors
  • For use in Memmert™ universal ovens
Specifications
Specifications
Secondary/Additional PT 100 Sensor (up to 3 can be attached in total)
Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108
Temperature Sensor
Load/Chamber Temperature Measurement
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Flexible Pt100 For Chamber Or Load, Display And Atmocontrol Compatible, For Un160Pa >