Learn More
Memmert™ Flexible Pt100 For Chamber Or Load, Display And Atmocontrol Compatible, For Un75Pa
Product Code. 16334507 Shop All Memmert Products
Click to view available options
For Use With (Application):
Load/Chamber Temperature Measurement
Unit Size:
Each
Product Code. 16334507
Brand: Memmert™ UN75paH8
Only null left
Add to basket
Add to basket
Includes
4-Pin Connector for Temperature Recording
Flexible temperature sensor for positioning in chamber or in load. Memmert™ PT100 Temperature Sensor is designed with a 4-pin connector for external temperature recording.
  • Can be positioned in chamber or in load
  • Includes 4-pin connector, according to NAMUR NE 28, for external temperature recording (load temperature) max. 3 sensors
  • For use in Memmert™ universal ovens
Specifications
Description Secondary/Additional PT 100 Sensor (up to 3 can be attached in total)
For Use With (Equipment) Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108
Type Temperature Sensor
For Use With (Application) Load/Chamber Temperature Measurement