Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF75
Flexible temperature sensor for positioning in chamber or in load.
Brand:  Memmert™ UF75H8
Product Code. 17529675
  • £197.00 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
Specifications
UF75 Oven
Temperature Sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UF75 > For Use With: UF75