missing translation for 'onlineSavingsMsg'
Learn More

3M™ Doodlebug™ Floor Edge Cleaning Kit

Convenient handle options provide versatile cleaning solutions. Unique hook and loop bottom holds 3M™ Doodlebug™ Pads, Sponges and Brushes without the use of adhesives or fasteners.

£250.00 - £288.00

Specifications

Product Type Cleaning Kit
Color Brown
For Use With (Application) Cleaning
For Use With (Equipment) Doodlebug™ Pads, Sponges and Brushes
View More Specs

Products 2
Product Code Brand Style Price Quantity & Availability  
Product Code Brand Style Price Quantity & Availability  
17128560
View Documents
3M™ Doodlebug™
7000002233
Handheld Pad Holder
£288.00
Pack of 4
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
 
17169350
View Documents
3M™ Doodlebug™
7100193869
Swivel Head Pad Holder (Universal Pole)
£250.00
6 sets
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
 
Description

Description

  • Ideal for maintenance of hard to reach areas
  • Efficient and versatile floor cleaning tool
  • Use the included white pad for polishing operations, and the brown pad for stripping
  • Pads are highly durable and easy to clean
Specifications

Specifications

Cleaning Kit
Brown
Cleaning
Doodlebug™ Pads, Sponges and Brushes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Doodlebug™ Floor Edge Cleaning Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.