missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scotch-Brite™ Floor Pads, Green
For the efficient, intermediate cleaning of heavily soiled floors with or without a coating. For use with single disc machines or automatic floor cleaning machines with slow speed.
£25.30 - £416.00
Specifications
Product Type | Floor Pad |
---|---|
Color | Green |
For Use With (Application) | Scrubbing |
For Use With (Equipment) | Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines |
Style | Round |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
17168800
|
Scotch-Brite™
7100159535 |
254 mm |
£25.30
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17198810
|
Scotch-Brite™
7100159890 |
280 mm |
£30.10
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17148820
|
Scotch-Brite™
7100159895 |
505 mm |
£86.90
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17138810
|
Scotch-Brite™
7100159883 |
530 mm |
£123.00
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17178800
|
Scotch-Brite™
7100159536 |
305 mm |
£35.00
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17138820
|
Scotch-Brite™
7100159894 |
330 mm |
£37.60
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17158810
|
Scotch-Brite™
7100159885 |
355 mm |
£42.35
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17198800
|
Scotch-Brite™
7100159879 |
380 mm |
£48.25
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17108810
|
Scotch-Brite™
7100159880 |
406 mm |
£54.30
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17118810
|
Scotch-Brite™
7100159881 |
432 mm |
£61.10
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17128810
|
Scotch-Brite™
7100159882 |
460 mm |
£416.00
Pack of 25 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17188810
|
Scotch-Brite™
7100159889 |
480 mm |
£92.85
Pack of 5 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
17277206
|
Scotch-Brite™
7100159882 |
460 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Floor Pad | |
Scrubbing | |
Round |
Green | |
Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines |