missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scotch-Brite™ Floor Pads, Red
Designed for everyday cleaning and light scrubbing to remove light soil, scuff marks, and black heel marks. These pads clean when damp and buff when dry. Open-web construction means this washable, reusable pad resists clogging and loading.
£37.40 - £197.00
Specifications
Product Type | Floor Pad |
---|---|
Color | Red |
For Use With (Application) | Everyday Cleaning, Light Scrubbing |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
17168850
|
Scotch-Brite™
7100182735 |
203 mm |
£60.15
Pack of 10 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17138840
|
Scotch-Brite™
7100182722 |
254 mm |
£37.40
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17128850
|
Scotch-Brite™
7100182731 |
480 mm |
£92.90
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17138850
|
Scotch-Brite™
7100182732 |
505 mm |
£105.00
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17148850
|
Scotch-Brite™
7100182733 |
530 mm |
£123.00
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17158850
|
Scotch-Brite™
7100182734 |
584 mm |
£197.00
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17148840
|
Scotch-Brite™
7100182723 |
280 mm |
£38.85
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17158840
|
Scotch-Brite™
7100182724 |
305 mm |
£45.30
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17168840
|
Scotch-Brite™
7100182725 |
330 mm |
£46.25
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17178840
|
Scotch-Brite™
7100182726 |
355 mm |
£50.90
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17188840
|
Scotch-Brite™
7100182727 |
380 mm |
£57.85
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17198840
|
Scotch-Brite™
7100182728 |
406 mm |
£65.20
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17108850
|
Scotch-Brite™
7100182729 |
432 mm |
£73.35
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
17118850
|
Scotch-Brite™
7100182730 |
460 mm |
£82.80
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
Description
- For everyday cleaning and light scrubbing
- Removes light soil, scuff marks, and black heel marks
- Cleans when damp and buffs when dry
- Conformable fibers provide maximum pad-to-floor contact
- Reversible, washable pad resists clogging and loading
- Consistent, proven performance with long floor pad life
Specifications
Floor Pad | |
Red | |
Everyday Cleaning, Light Scrubbing |