Learn More
Cole-Parmer™ Stuart™ Florentine Flask For Stuart™ Rotary Evaporators
Evaporating flask used with Stuart™ Rotary Evaporators.
Brand:  Cole-Parmer™ 10096-55
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 12477732
  • £167.80 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket
Description
Description
  • For use with Stuart™ rotary evaporators
  • Flat bottom
  • 29/32 socket
Specifications
Specifications
Evaporating flask
Glass
Flat
1 L
29/32