missing translation for 'onlineSavingsMsg'
Learn More
Florisil™, 30-60 mesh, for column chromatography Product Code.: 15545266

Florisil™, 30-60 mesh, for column chromatography

Product Code. 15545266
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 15545266

Brand: Thermo Scientific Acros 457735000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1343-88-0 | MgO3Si | 100.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1343-88-0
Quantity 500 g
Molecular Formula MgO3Si
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Solubility Insoluble in water
Molecular Weight (g/mol) 100.39
Formula Weight 100.39
Chemical Name or Material Florisil™

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Florisil™, 30-60 mesh, for column chromatography >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.