missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Florisil, 60-100 mesh, for column chromatography, Thermo Scientific Chemicals
Florisil, MgO3Si, CAS Number-1343-88-0, 1kg, White, 01,393; 09,223, 100.39, ZADYMNAVLSWLEQ-UHFFFAOYSA-N, 10, 5509, MFCD00078249, 60-100 mesh, for column chromatography, Glass bottle
£73.16 - £786.92
Chemical Identifiers
CAS | 1343-88-0 |
---|---|
Molecular Formula | MgO3Si |
Molecular Weight (g/mol) | 100.39 |
MDL Number | MFCD00078249 |
InChI Key | ZADYMNAVLSWLEQ-UHFFFAOYSA-N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10358580
|
Thermo Scientific Acros
205452500 |
250 g | Glass Bottle |
£73.16
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174830
|
Thermo Scientific Acros
205455000 |
500 g | Glass Bottle |
£118.59
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10793011
|
Thermo Scientific Acros
205450010 |
1 kg | Glass Bottle |
£203.01
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10512904
|
Thermo Scientific Acros
205450050 |
5 kg | Plastic Drum |
£786.92
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatographyChemical Identifiers
1343-88-0 | |
100.39 | |
ZADYMNAVLSWLEQ-UHFFFAOYSA-N |
MgO3Si | |
MFCD00078249 |
Specifications
White | |
1343-88-0 | |
MFCD00078249 | |
10, 5509 | |
ZADYMNAVLSWLEQ-UHFFFAOYSA-N | |
100.39 |
Fine Powder | |
MgO3Si | |
01,393; 09,223 | |
Particle size analysis (typical values): 60 mesh: 2.0%, 70 mesh: 20.0%, 80 mesh: 41.0%, 100 mesh: 30.0%, 140 mesh: 6.9% | |
100.39 | |
Florisil™, 60-100 mesh, for column chromatography |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title