missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, Activated at 675°C, 60 to 100 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

£460.00 - £1510.00

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15508124
View Documents
Avantor J.T.Baker
3372-07
500 g Wide Mouth Amber Glass Bottle
£460.00
500g
Please to purchase this item. Need a web account? Register with us today!
15518124
View Documents
Avantor J.T.Baker
3372-08
2 kg Poly Pail
£1510.00
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    Suitable for Use in Chromatographic Cleanup of Pesticide Residues
Specifications

Chemical Identifiers

1343-88-0
MgO3Si
100.39
ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

1343-88-0
60 to 100 Mesh
100.39
BAKER ANALYZED™ Reagent
MgO3Si
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
1L = 2.51kg
Florisil™, Activated at 675°C
Videos
SDS
Documents

Documents

Product Certifications