Learn More
Florisil™, for Chromatography, 60-100 Mesh, Fisher Chemical
Brand: Fisher Chemical F/1170/50
116.40 GBP valid until 2024-07-31
Use promo code "21824" to get your promotional price.
Code : 21
Additional Details : CAS Number : 1343-88-0 Weight : 0.25000kg
Specifications
Plastic Powder Jar | |
250 g |