Learn More
Cytiva Flow Cell
UV flow cell
Brand: Cytiva 18-1128-24
Code : GE
Additional Details : Weight : 0.03200kg
Description
UV Flow CellSpecifications
Flow Cell | |
6 μL | |
ETFE, Glass, Titanium, Fluorcarbon Rubber and Encapsulated Silicone O-ring |