Learn More
OHAUS™ Flow Control Clamp
OHAUS™ Flow Control Clamp in a variety of styles, materials, and sizes.
Brand:  OHAUS™ 30392210
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15742259
  • £23.50 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Designed to securely grip and position laboratory apparatus
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
  • Available as single or dual-adjust in zinc or stainless steel
Specifications
Specifications
Multi-purpose Clamp
Clamp, Flow Control, Hosecock, CLF-HKRGZ
2.44 in.
0 to 17 mm
37.00 mm
Nickel-plated
62.000 mm
37 mm
1.46 in.
Hosecock