Learn More
OHAUS™ Flow Control Clamp
OHAUS™ Flow Control Clamp in a variety of styles, materials, and sizes.
Brand: OHAUS™ 30392221
This item is not returnable.
View return policy
Description
- Designed to securely grip and position laboratory apparatus
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Available as single or dual-adjust in zinc or stainless steel
Specifications
Multi-purpose Clamp | |
Clamp, Flow Control, Hosecock, CLF-HKRGS | |
2.44 in. | |
0 to 17 mm | |
37.00 mm |
Stainless Steel | |
62.000 mm | |
37 mm | |
1.46 in. | |
Hosecock |