Learn More
IKA™ FLOW System
Flow chemistry made easy and affordable with IKA FLOW. Its secret: Unlike other commercially available flow chemistry equipment, you just purchase the necessary part of the flow chemistry apparatus.
Brand:  IKA™ 0020100742
Additional Details : Weight : 5.62000kg
Product Code. 17584066
  • £6325.00 / Each
Estimated Shipment: 05-11-2024
to see stock.
Add to basket
Description
Description
  • The portable IKA FLOW is an attachment that forms a flow chemistry reactor in combination with your IKA magnetic stirrer RCT digital. In addition, it has been designed to be used with any fume hood gas source (with a 6 mm pressure tubing and an adaptor to typical fume hood gas supplies) such as compressed air or nitrogen.
  • IKA FLOW's scope of delivery includes in addition to the attachment, a glass bottle, a mixing chip, support screws, three on-and-off valves, a heating block, four disc reactors, and a labeled tubing set.
  • Pair IKA FLOW with the RCT digital stirrer hotplate.
  • Scope of delivery
    • IKA FLOW System
    • Glass Bottle
    • Heating Block
    • CDR.PP standard (4 pcs.)
    • Capillary Set
    • Tubing set
Specifications
Specifications
500 mL
2.7 kg
Glass Bottle, Mixing Chip, Support Screws, Three Valves, Heating Block, Four Disc Reactors, Labeled Tubing Set
304 Stainless Steel, Nylon (PA)
IKA Magnetic Stirrer RCT Digital