missing translation for 'onlineSavingsMsg'
Learn More

Floxuridine, MedChemExpress

Product Code. 16411376
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16411376

Brand: Medchem Express HYB0097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis. Floxuridine has antiviral effects against HSV and CMV.

Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis. Floxuridine has antiviral effects against HSV and CMV.

Specifications

Chemical Name or Material Floxuridine
CAS 50-91-9
Color White
Purity Grade Notes Research
Molecular Formula C9H11FN2O5
Quantity 200 mg
Synonym 5-Fluorouracil 2'-deoxyriboside
Solubility Information DMSO : 250 mg/mL (1015.48 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (203.10 mM)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C(F)=C2)=O)=O)O1)O
Molecular Weight (g/mol) 246.19
Formula Weight 246.19
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.