missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Floxuridine, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Description
Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis. Floxuridine has antiviral effects against HSV and CMV.
Specifications
Specifications
| Chemical Name or Material | Floxuridine |
| CAS | 50-91-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C9H11FN2O5 |
| Quantity | 100 mg |
| Synonym | 5-Fluorouracil 2'-deoxyriboside |
| Solubility Information | DMSO : 250 mg/mL (1015.48 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (203.10 mM) |
| SMILES | OC[C@@H]1[C@H](C[C@H](N2C(NC(C(F)=C2)=O)=O)O1)O |
| Molecular Weight (g/mol) | 246.19 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction