missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flt-3 Ligand Human ProQuantum Immunoassay Kit
GREENER_CHOICE

Invitrogen™ ProQuantum™ Immunoassay Kit measures proteins with no wash steps, small sample consumption, high sensitivity, and large dynamic range on a qPCR instrument

£648.00 - £2895.00

Specifications

Accession Number P49771
Assay Range 0.064 to 25000 pg/mL
Assay Sensitivity <0.05 pg/mL
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15910473
GREENER_CHOICE
View Documents Promotion Details
Invitrogen™
A35581
96 Tests
£648.00
Each
In Stock
to see stock.
Add to basket
 
17183099
GREENER_CHOICE
View Documents Promotion Details
Invitrogen™
A355815
5 x 96 Tests
£2895.00
Each
Estimated Shipment: 28-11-2024
to see stock.
Add to basket
 
Description

Description

  • High-sensitivity: Detect low levels of proteins with greater sensitivity than traditional methods like ELISA
  • Small sample consumption: Use 2 to 5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
  • Fast, easy workflow: No wash steps, 2 hours from sample to answer
  • Broad dynamic range: 5 logs minimizes sample dilutions to ensure falling within the range
  • No proprietary instrument to purchase: Runs on any real-time PCR instrument
  • ProQuantum software available: Robust data analysis and statistical group wise comparison
Spezifikation

Spezifikation

P49771
<0.05 pg/mL
Proximity-based Immunoassay
Human
qPCR Instruments
1 hr.
FAM™
Serum,2-5μL; Supernatant,2-5μL; Plasma,2-5μL
Human
2 hr.
0.064 to 25000 pg/mL
Taqman™ Real-time PCR
ProQuantum
Protein quantitation
2323
Antibody-conjugate A, Antibody-conjugate B, Antibody-conjugate dilution buffer, Assay dilution buffer, Ligase, Master Mix, Standard
RUO
Frozen at -20°C
30 min.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen