missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole, Tocris Bioscience™

Product Code. 18742664
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Triazole antifungal agent

Triazole antifungal agent. Effective against Candida strains in vitro and in vivo.

Specifications

CAS 86386-73-4
Molecular Formula C13H12F2N6O
Quantity 50 mg
Synonym fluconazole, diflucan, triflucan, elazor, biozolene, biocanol, fungata, fluconazol, fluconazolum, flucazol
InChI Key RFHAOTPXVQNOHP-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)F)C(CN2C=NC=N2)(CN3C=NC=N3)O
IUPAC Name 2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
Molecular Weight (g/mol) 306.277
PubChem CID 3365
ChEBI CHEBI:46081
Formula Weight 306.27
Percent Purity >98%
Chemical Name or Material Fluconazole
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.