missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fludarabine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Purine analog that inhibits DNA synthesis. Exhibits antiproliferative activity (IC50 = 1.54 μM in RPMI cells) and triggers apoptosis through increasing Bax and decreasing Bid, XIAP and survivin expression. Inhibits cytokine-induced activation of STAT1 and STAT1-dependent gene transcription in lymphocytes. Also displays anticancer activity against hematological malignancies in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 285.23 |
Percent Purity | >99% |
Chemical Name or Material | Fludarabine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'