missing translation for 'onlineSavingsMsg'
Learn More

Flufenamic acid, Tocris Bioscience™

Product Code. 18742784
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium-activated chloride channel blocker; NSAID. Also activates TRPC6

Nonsteroidal anti-inflammatory drug (NSAID). Inhibits calcium-activated chloride channels (CaCCs). Also increases currents through TRPC6 channels and inhibits currents through TRPC3 and TRPC7 channels.

Specifications

CAS 530-78-9
Molecular Formula C14H10F3NO2
Quantity 50 mg
Synonym flufenamic acid, fluphenamic acid, nichisedan, achless, arlef, flufacid, fullsafe, lanceat, paraflu, plostene
InChI Key LPEPZBJOKDYZAD-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(=O)O)NC2=CC=CC(=C2)C(F)(F)F
IUPAC Name 2-[3-(trifluoromethyl)anilino]benzoic acid
Molecular Weight (g/mol) 281.234
PubChem CID 3371
ChEBI CHEBI:42638
Formula Weight 281.23
Percent Purity >99%
Chemical Name or Material Flufenamic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'