Learn More
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Product Code. p-4523848
Click to view available options
Quantity:
10/Pk.
Format:
SB
Media Type:
Fluid Thioglycollate Medium
Manufacturing Origin:
United States
Unit Size:
Pack of 10
Product Code. 17799438
Brand: Thermo Scientific™ R112641
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
  • For sterility testing according to USP (United States Pharmacopeia)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, USP.
Format SB
Manufacturing Origin United States
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
Quantity 10/Pk.