Learn More
Thermo Scientific™ Fluid Transfer Assembly
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
POA
Products 6
Product Code Brand Tubing Style Price Quantity & Availability  
Product Code Brand Tubing Style Price Quantity & Availability  
15929916
View Documents
Thermo Scientific™
SS00042-I
91.4cm of 12.7cm ID Silicone Tubing with CPC Steam-Thru II and ReadyMate DAC Connections
POA
25 units
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15969916
View Documents
Thermo Scientific™
SS00062-I
91.4cm of 12.7cm ID Silicone Tubing with 3.8cm Sanitary and 1.3cm MPX Body Connections
POA
25 units
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15949916
View Documents
Thermo Scientific™
SS00044-I
91.4cm of 12.7cm ID Silicone Tubing with Lynx ST and ReadyMate DAC Connections
POA
25 units
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15979916
View Documents
Thermo Scientific™
SS00063-I
91.4cm of 12.7cm ID Silicone Tubing with 1.9cm Sanitary and 1.3cm MPX Body Connections
POA
25 units
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15989916
View Documents
Thermo Scientific™
SS00064-I
91.4cm of 12.7cm ID Silicone Tubing with 3.8cm Sanitary and 1.3cm MPX insert Connections
POA
25 units
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15999916
View Documents
Thermo Scientific™
SS00065-I
91.4cm of 12.7cm ID Silicone Tubing with 1.9cm Sanitary and 1.3cm MPX insert Connections
POA
25 units
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.
  • End treatments facilitate simple interconnection with existing process and equipment
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality