missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom Fluido 2 Microplate Washer for 96 Well Plates with 2 Liquid Lines
Description
- Flexible microplate washer for 96 or 384-well plates
- Wide choice of washing options and programs providing maximum application options
- Low noise, pressure-free aspiration and dispensing pumps
- Auto rinse function preventing liquid path clogging
Specifications
Specifications
| Connections | USB 2.0 (USB A to B cable) |
| Description | Fluido 2 Microplate Washer for 96 Well Plates with 2 Liquid Lines |
| Bottles | 2 |
| Precision | 0.25% at 1.0 OD (at 450 nm) |
| Dispense Volume | 50 to 2000 μL |
| Plate Type | 96-Well Plates |
| Residual Volume | ±1 μL |
| Weight (Metric) | 6.2 kg |
| Display | LCD |
| For Use With (Application) | ELISA |
| Show More |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
