missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flumazenil, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Beschreibung
Benzodiazepine antagonist, non-selective for α1, α2, α3 or α5-containing GABAA receptors. Centrally active upon systemic administration in vivo. Also inhibits repressor element-1 silencing transcription factor (REST) by targeting degradation.
Spezifikation
Spezifikation
Quantity | 50 mg |
Formula Weight | 303.29 |
Percent Purity | >99% |
Chemical Name or Material | Flumazenil |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'