missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluo-4 Direct™ Calcium Assay Kit

Product Code. p-7075900
Click to view available options
Quantity:
10 x 10 mL
1 x 100 mL
1 x 1 L
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10297783

Brand: Molecular Probes™ F10471

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fluo-4 Direct™ Calcium Assay Kit

The Fluo-4 Direct™ Calcium Assay Kit was formulated to provide a homogeneous fluorescent calcium assay that:
1. Easily loads into cells
2. Achieves a large assay window and
3. Suppresses background fluorescence generated from the calcium indicator in complete media with little-to-no impact on the specific cellular fluorescence

This advanced formulation allows the assay to be run in a simple addition only format in the presence of serum-containing media. Fluo-4 Direct™ is the third addition to the Fluo-4 family of calcium detection reagents. Fluo-4 AM and Fluo-4 NW both require media removal before assay detection, but Fluo-4 NW adds convenience by including the PowerLoad™ reagent for ease in cell loading. The Fluo-4 Direct™ Calcium Assay Kit is similar to Fluo-4 NW in that it is formulated with PowerLoad™ for ease in cell loading, but unique in that it can be used in the presence of complete culture media and will efficiently suppress background fluorescence without sacrificing the specific cellular fluorescence generated in the assay.
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage
  • 10 × 10 ml Fluo-4 Direct™ Calcium Assay Reagent (Component A)
  • 2 × 77 mg Probenecid (Component B)
  • 200 ml Fluo-4 Direct™ Calcium Assay Buffer (Component C)
  • Store at ≤-20°C. Dessicate and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Assay
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product Type Calcium Assay Kit
Dye Type Fluorescent Dye-Based
Emission Visible
Form Powder
Modification Chemical
Product Line Molecular Probes
Quantity 10 x 10 mL
Shipping Condition Room Temperature
Sub Cellular Localization Nucleus, Organelles, Cytoplasm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.