missing translation for 'onlineSavingsMsg'
Learn More

Fluorene, 98+%

Product Code. 10437750 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10437750

Brand: Thermo Scientific Acros 156135000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-73-7 | C13H10 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-73-7
Molecular Formula C13H10
Molecular Weight (g/mol) 166.22
MDL Number MFCD00001111
InChI Key NIHNNTQXNPWCJQ-UHFFFAOYSA-N
Synonym fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
PubChem CID 6853
ChEBI CHEBI:28266
IUPAC Name 9H-fluorene
SMILES C1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 112.0°C to 116.0°C
Color Brown to White
Density 1.2000g/mL
Boiling Point 298.0°C
Flash Point 151°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 05, 625
Merck Index 15, 4187
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and carbon disu,freely soluble in glacial acetic acid
Formula Weight 166.22
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Fluorene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Fluorene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.