missing translation for 'onlineSavingsMsg'
Learn More

Fluorescamine, pure, Thermo Scientific Chemicals

Product Code. 10153730
Click to view available options
Quantity:
25 mg
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38183-12-9
Molecular Formula C17H10O4
Molecular Weight (g/mol) 278.26
MDL Number MFCD00005928
InChI Key ZFKJVJIDPQDDFY-UHFFFAOYNA-N
PubChem CID 37927
IUPAC Name 4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
SMILES O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 155.0°C to 157.0°C
Solubility Information Solubility in water: .. Other solubilities: soluble in diethyl ether
Infrared Spectrum Authentic
Formula Weight 278.26
Grade Pure
Packaging Glass bottle
Specific Extinction 900 (235nm, ether) min.
Color Yellow
Quantity 100 mg
Physical Form Powder
Chemical Name or Material Fluorescamine
Show More Show Less
compliance-icons
Product Identifier
  • Fluorescamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'