missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluorescein-5-Isothiocyanate (FITC 'Isomer I')

Fluorescein-5-Isothiocyanate (FITC 'Isomer I')

Brand:  Molecular Probes™ F1906

120.84 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11574786

  • £159.00 / 100mg
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 x 10 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
FITC (Fluorescein)
Fluorescein-5-Isothiocyanate
Amine
Isothiocyanate
Classic Dyes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.