missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Fluorescein diacetate, 97%, pure Product Code.: 10067432

Thermo Scientific Chemicals Fluorescein diacetate, 97%, pure

Product Code. 10067432
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10067432

Brand: Thermo Scientific Chemicals 191660250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-09-8
Molecular Formula C24H16O7
Molecular Weight (g/mol) 416.39
MDL Number MFCD00005062
InChI Key CHADEQDQBURGHL-UHFFFAOYSA-N
Synonym fluorescein diacetate, 3,6-diacetoxyfluoran, diacetylfluorescein, di-o-acetylfluorescein, fluorescein, diacetate, 3',6'-diacetylfluorescein, unii-yl39r93pre, yl39r93pre, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 3',6'-bis acetyloxy, 3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-3',6'-diyl diacetateShow MoreShow Less
PubChem CID 65047
IUPAC Name (6'-acetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)OC(=O)C)C5=CC=CC=C5C(=O)O3

Specifications

Melting Point 200.0°C to 205.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 19,227
Formula Weight 416.39
Percent Purity 97%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein diacetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461957 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number191660250,191660050,191661000
Lot NumberA0461263 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number191660250,191660050,191661000
Lot NumberA0454971 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number191660250,191660050,191661000
Lot NumberA0448193 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number191660250,191660050,191661000
Lot NumberA0404012 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number191660250,191660050,191661000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Fluorescein diacetate, 97%, pure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.