missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Sodium, Pure, C.I.45350, Fisher Chemical™

Product Code. 10090160
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorescein Sodium, Pure, C.I.45350

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Melting Point 320°C
Color Brown-Red
Physical Form Powder/Solid
Quantity 100 g
Formula Weight 376.27g/mol
Packaging Glass Jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'