missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Sodium Salt, indicator, 98.5 to 100.5%, Honeywell Fluka™

Product Code. 15602940
Click to view available options
Quantity:
100 g
1 kg
4 x 1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, 98.5-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Melting Point >300°C
UN Number NONH for all modes of transport
Beilstein 3833041
Formula Weight 376.27g/mol
Percent Purity 98.5 to 100.5%
Grade Indicator
Packaging Glass Bottle
Color Red
Quantity 4 x 1 kg
Physical Form Powder
Chemical Name or Material Fluorescein Sodium Salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'