missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Sodium Salt, indicator, 98.5 to 100.5%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
4 x 1 kg
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
UN Number | NONH for all modes of transport |
Beilstein | 3833041 |
Formula Weight | 376.27g/mol |
Percent Purity | 98.5 to 100.5% |
Grade | Indicator |
Packaging | Glass Bottle |
Color | Red |
Quantity | 100 g |
Physical Form | Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'