Learn More
Agilent BioTek Fluorescence Filter
Fluorescence Filter for use with Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Brand:  Agilent BioTek 7082223
Code : NEW
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10566825
  • £680.00 / Each
Estimated Shipment: 25-07-2024
to see stock.
Description
Description
Fluorescence Filter for use with Agilent BioTek Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Specifications
Specifications
Fluorescence Filter
Cytation 3, Cytation 5, Cytation Multi-Mode, FLx800, Synergy 2, Synergy H1, Synergy HTX Multi-Mode Readers
530/25 nm