missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescent Compatible Sample Buffer (4X, non-reducing)

Invitrogen Fluorescent Compatible Sample Buffer is for use in SDS-polyacrylamide gel electrophoresis (SDS-PAGE).

Brand:  Invitrogen™ LC2570

38.89 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 15940783

  • £63.75 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

It is optimized for use in fluorescent western blotting as it does not interfere in fluorescent channels, unlike other standard sample buffers. Fluorescent Compatible Sample Buffer is non-reducing but may also be used under denaturing conditions.

Non-reducing—ready-to-use for non-reducing SDS-PAGE, or the preferred type and amount of reducing agent (e.g., DTT) can be added to produce reducing conditions
Lane marker dyes—visible markers of the dye front
Convenient—stable at room temperature, enabling storage at the bench where electrophoresis is performed
Concentrated—4X formulation provides more versatility than traditional 2X buffers
Ideal for fluorescent detection—formulated to prevent auto fluorescence during fluorescent detection

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
4X
Tricine, Tris-Glycine, SDS-PAGE, Bis-Tris, Bis-Tris Plus, Tris-Acetate
SDS-PAGE Gels
Room Temperature
Sample Loading Buffers
Fluorescence
Fluorescent Compatible Sample Buffer
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.