missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorexon

Product Code. 10154161
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-15-0
Molecular Formula C30H26N2O13
Molecular Weight (g/mol) 622.539
MDL Number MFCD00005049
InChI Key DEGAKNSWVGKMLS-UHFFFAOYSA-N
Synonym Bis[N, N-bis(carboxymethyl)aminomethyl]fluorescein, Calcein, Fluorescein Complexon
PubChem CID 65079
ChEBI CHEBI:51903
IUPAC Name 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Infrared Spectrum Authentic
Formula Weight 622.54
Packaging Glass bottle
Suitability Suitable for calcium determination
UV Spectrometry Complies
Color Brown/Orange to Orange/Yellow
Quantity 25 g
Physical Form Powder
Chemical Name or Material Fluorexon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.