missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoride ISE Standard Solution, Mettler Toledo™
114.44 GBP valid until 2024-12-31
Use promo code "23284" to get your promotional price.
Use promo code "23284" to get your promotional price.
Click to view available options
Quantity:
500 mL
Specifications
Description | ISE standard F 1000 mg/L, 500mL |
Packaging | 500 mL |
Quantity | 500 mL |
Color | Colorless |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'