missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoride solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£50.50 - £119.00
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | H2O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12982115
|
Fisher Chemical
A 017 100 |
100 mL |
£50.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12992115
|
Fisher Chemical
A 017 250 |
250 mL |
£102.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12902125
|
Fisher Chemical
A 017 500 |
500 mL |
£119.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
1g/L | |
H2O |