missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ Fluorimeter Interference Filter

Requires a much more narrow range of wavelengths. Jenway™ Fluorimeter Interference Filter allows a specified wavelength and light to pass through to the sample.

Brand:  Jenway™ 10089-79

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11464879

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • For use with Jenway™ Fluorimeters
  • Normally used as the emission filters where a much narrower range of wavelengths is required
  • Allow the specified wavelength and light ±5nm of this through to the sample.
Specifications

Specifications

Interference Filter
405 nm
Jenway Fluorimeters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers