Learn More
Thermo Scientific™ Fluoro-Max Fluorescent Streptavidin-Coated Particles
Simplify the binding process with a universal binding reagent.
£244.00 - £12655.00
Specifications
Additive | 0.05% Sodium Azide |
---|---|
Binding Property | Streptavidin |
Concentration | 1% solids |
Content And Storage | Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. |
Contents | Fluorescent red dyed polystyrene microspheres in water |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11864992
|
Thermo Scientific™
29470701010350 |
100 mL |
£12655.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11874992
|
Thermo Scientific™
29470701011150 |
1 mL |
£244.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11854992
|
Thermo Scientific™
29470701010150 |
5 mL |
£855.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Simplify the binding process with a universal binding reagent by using Thermo Scientific™ Fluoro-Max Fluorescent Streptavidin-Coated Particles in a variety of applications such as clinical diagnostics, immuno/histological studies and research. With an extremely broad Stokes shift, europium chelate particles help prevent nonspecific fluorescence interference. They are also excellent for quantitative membrane-based rapid assays, heterogeneous assays, luminescent assays, research applications, phagocytosis studies, cell surface markers and pore size determination.
When excited with UV light at its maximal absorbance wavelength of 333 nm, europium chelate particles emit long-lived luminescence at 613 nm, with a lifetime of approximately 0.5 milliseconds, which is about 10,000 to 100,000 times longer than the lifetimes of most fluorophores. This extremely long lifetime allows the application of europium particles to time-resolved fluorescence and can reduce the detection limits of fluorescence assays many orders of magnitude by elimination of background interference from relatively short-lived matrix fluorescence.
- Made by dyeing Thermo Scientific™ OptiLink™ Carboxylate-Modified particles with europium chelate
- Specifically designed for membrane or automated fluorometric-based applications
- Available in standard 0.3 μM diameter
- Dyed internally to prevent dye leaching and to assure maximum surface immunoreactivity
Fluoro-Max Fluorescent Benefits
- Bind ligands without dye interference
- Dye-free surface for coupling
- High protein binding capacity
- Readily adsorbs proteins
- Optimize assays by controlling sensitivity, specificity and stability
- Optimized acid content
Recommended for:
- Clinical diagnostics
- Molecular biology
- Lateral flow, turbidimetric and slide aggulation assays
Specifications
0.05% Sodium Azide | |
1% solids | |
Fluorescent red dyed polystyrene microspheres in water | |
Immunohistological Studies | |
Fluorescent Streptavidin-Coated Particles | |
Package Insert Sheet containing Certicate of Analysis | |
1.06g/cm³ | |
Streptavidin | |
0.3 μm | |
0.3 μm |
Streptavidin | |
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. | |
White | |
Polystyrene or Polystyrene with Coploymer Grafted Surface Coated with Streptavidin | |
Europium Chelate | |
Biotin binding of ∼1000pmol/mg | |
Product stable for a minimum of 24 months | |
<5% CV | |
0.3 μm |