missing translation for 'onlineSavingsMsg'
Learn More

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Product Code. 15461756
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164298-23-1
Molecular Formula C5H12F7N2P
Molecular Weight (g/mol) 264.128
MDL Number MFCD02684443
InChI Key ZAVXOOLKAGPJPI-UHFFFAOYSA-N
Synonym tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, n-dimethylamino fluoromethylene-n-methylmethanaminium hexafluorophosphate v, tetramethyl fluoroformamidium hexafluoro phosphate, n,n,n',n'-tetramethylfluoroformamidinium hexafluorophosphate, tetramethylfluoroformamidinium hexafluorophosphate, tffh, fluoro-n,n,n',n'-tetramethylformamidinium hexafluorophosphate, c5h12fn2.f6p, acmc-209dqu, tetramethyl fluoroformamidium hexafluorophosphate
PubChem CID 2774761
IUPAC Name [dimethylamino(fluoro)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)F.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 104°C to 111°C
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 264.12
Percent Purity 98%
Chemical Name or Material Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'