Learn More
Cytiva Fluoro Plastics Fluoro Rubber Glass Superloop
Introduce large sample volumes into a pressurised fluid LC system. Cytiva Fluoro Plastics Fluoro Rubber Glass Superloop is used together with the injection valve MV-7, INV-907, V9-Inj, or V9H-Inj and replaces a simple sample loop.
Brand: Cytiva 19-7850-01
Code : GX
Additional Details : Weight : 0.49900kg
Description
- Superloop can be used in conjunction with 7-port valves.
- The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection.
- They may also be used to inject pulses of a specific buffer solution.
- Flexible: inject large or small sample volumes.
- High reproducibility and recovery.
- No gradient dilution with internal movable seal.
Specifications
Superloop | |
Fluoro Plastics, Fluoro Rubber, Glass | |
50 mL |