Learn More
Fluorobenzene, 99%, Thermo Scientific Chemicals
£17.69 - £153.77
Chemical Identifiers
CAS | 462-06-6 |
---|---|
Molecular Formula | C6H5F |
Molecular Weight (g/mol) | 96.10 |
MDL Number | MFCD00000280 |
InChI Key | PYLWMHQQBFSUBP-UHFFFAOYSA-N |
Synonym | benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene |
PubChem CID | 10008 |
ChEBI | CHEBI:5115 |
IUPAC Name | fluorobenzene |
SMILES | FC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10733671
|
Thermo Scientific Acros
119341000 |
100 g | Glass bottle |
£17.69
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317040
|
Thermo Scientific
119345000 |
500 g | Glass bottle |
£53.37
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904521
|
Thermo Scientific Acros
119340025 |
2.5 kg | Glass bottle |
£153.77
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
462-06-6 | |
96.10 | |
PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
10008 | |
fluorobenzene |
C6H5F | |
MFCD00000280 | |
benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene | |
CHEBI:5115 | |
FC1=CC=CC=C1 |
Specifications
462-06-6 | |
Colorless | |
85.0°C | |
Authentic | |
C6H5F | |
1.4643 to 1.4663 | |
05,IV,632 | |
1.02 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol and water | |
FC1=CC=CC=C1 | |
96.10 | |
CHEBI:5115 | |
99% | |
Fluorobenzene |
-42.0°C | |
1.0200g/mL | |
−15°C | |
98.5% min. (GC) | |
C6H5F | |
MFCD00000280 | |
15,4201 | |
benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene | |
PYLWMHQQBFSUBP-UHFFFAOYSA-N | |
fluorobenzene | |
10008 | |
96.1 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 207-321-7
- Fluorobenzene
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only