missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoroboric Acid, Technical, 50% Solution, Fisher Chemical
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
Specifications
Specifications
Color | Colorless to Yellow |
CAS | 16872-11-0 |
Melting Point | -90°C |
Boiling Point | 130°C |
Packaging | HDPE plastic bottle |
Vapor Pressure | 5.1mmHg at 20°C |
Quantity | 500 mL |
Molecular Formula | BF4H |
Physical Form | Liquid |
MDL Number | 11345 |
Show More |
Safety and Handling


Product Identifier
- Fluoroboric acid, 50 wt% solution in water
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fluoroboric Acid, Technical, 50% Solution, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction